Would you like to log in again?
Labclean™ Cleanroom Wipes, 100% Polyester, United Scientific Supplies
Supplier: United Scientific Supplies
PWIPE4X4 PWIPE12X12 PWIPE9X9
470339-168EA 34.09 USD InStock
470339-168 470340-094 470339-504
Labclean™ Cleanroom Wipes, 100% Polyester, United Scientific Supplies
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polyester wipes for cleanroom environments.
  • 100% virgin, continuous filament, double-knit polyester fiber
  • Laser cut sealed edge for reduced fiber contamination
  • Moderate absorbency and abrasion resistance
  • Low levels of particulate and extractable counts
Polyester wipes are 100% virgin, continuous-filament polyester in a double-knit, no-run interlock design with a laser cut sealed edge. Recommended for use in cleanroom critical environments and a wide variety of applications including cleaning medical devices, electronics displays, instrument panels, semiconductors, and microelectronics products. Laundered and packaged in a cleanroom. Vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with an inner bag label. Available in three convenient sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty