Would you like to log in again?
Ampere's Rule Apparatus
Supplier: United Scientific Supplies
AMPR01
470339-572EA 78.15 USD InStock
470339-572
Ampere's Rule Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate the right hand rule for the magnetic field around a current carrying wire.
  • Designed to help students study the magnetic field around a wire
  • Contains heavy brass wire with terminals, arranged on a clear plastic base
  • One 45 mm compass and six 16 mm compasses also included
  • Comes with activity guide
This apparatus allows students to study the magnetic field around a wire. A heavy brass wire with terminals is arranged on a clear plastic base. Also included are one each 45 mm compass, six each 16mm compasses, and an activity guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty