Would you like to log in again?
Hydrogen and Helium Spectrum Tube
Supplier: United Scientific Supplies
SPT2HH1
470339-594EA 42.7 USD InStock
470339-594
Hydrogen and Helium Spectrum Tube
Educational Materials Physics Educational Materials Optics Learning Activities
Illustrate how spectral patterns can be analyzed from a combination of gases.
  • Ideal for classroom demonstrations
  • Includes an activity guide
During most of their life, stars are mostly composed of hydrogen and generate energy by converting it to helium. Although other elements are present to give individual stars their 'fingerprint' the lines of hydrogen and helium are visible in stellar spectra, as in this tube.
The glass tubes are 25 cm long with the capillary portion about 5 cm long. An electrode is sealed in each end and attached to a metal end cap with a loop for convenient hookup to wire connections. Each spectrum tube includes an activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty