Would you like to log in again?
Large U-Shaped Electromagnet
Supplier: United Scientific Supplies
EMGT01
470339-632EA 30.35 USD InStock
470339-632
Large U-Shaped Electromagnet
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate the power of an electromagnet.
  • Classroom electromagnet lifts up to 3.5 kg
  • Operates on 1.5 to 6 V DC
Operating on 1.5 to 6 V DC, this classroom electromagnet lifts up to 3.5 kg. A current direction indicator is included for the study of the magnetic field.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty