Would you like to log in again?
United Scientific™ Inclined Plane With Pulley
Supplier: United Scientific Supplies
INPL01
470339-638EA 98 USD InStock
470339-638
United Scientific™ Inclined Plane With Pulley
Educational Materials Physics Educational Materials Mechanics Learning Activities
Investigate forces on an incline plane.
  • Durable steel inclined plane (4W×23" L) for classroom demonstrations
  • Includes a graduated protractor and an adjustable pulley
  • Includes a 14 oz. roller that can be used as a vehicle on the plane, or as a counterweight
  • Includes a pan and cord for masses, as well as an activity guide
This versatile and tough steel inclined plane includes a graduated protractor and an adjustable pulley.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty