Would you like to log in again?
Bottle Top Dispenser, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BTDR-1 BTDR-5 BTDR-4 BTDR-6 BTDR-2 BTDR-3
470339-962EA 435.75 USD InStock
470339-962 470339-216 470339-642 470339-086 470340-084 470339-180
Bottle Top Dispenser, United Scientific Supplies
Bottle Top Dispensers
Precise dispensing for laboratory solutions.
  • Includes adapters to fit a wide range of bottle sizes: 28, 30, 32, 36, 40 and 45 mm
  • All wetted parts made from PTFE and borosilicate glass for maximum chemical resistance
  • Not suitable for use with hydrogen fluoride
  • Individually calibrated and certified in an ISO 17025 accredited laboratory in accordance with ISO 8655 standards
  • Calibration certificate included
Five adapters are included with each unit for use with a wide range of bottle sizes. All wetted parts are made from PTFE and borosilicate glass for maximum chemical resistance. Suitable for all laboratory applications except hydrogen fluoride. Special precautions must be taken for use, cleaning, and storage when working with concentrated fuming acids.
A factory calibration certificate is provided. Easy to disassemble, and can be recalibrated in the lab.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty