Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crucibles, Wide Form with Cover, Porcelain, United Scientific Supplies

Supplier: United Scientific Supplies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

JCL150 JCL030 JCL050 JCL100 JCL250 JCL020
470338-944EA 90.7 USD InStock
470338-944 470340-158 470339-844 470340-082 470339-652 470338-938
Crucibles, Wide Form with Cover, Porcelain, United Scientific Supplies
Crucibles
Wide form crucibles include covers. Crucibles are glazed on inner and outer surfaces except for the outside bottom and rim.

  • Autoclavable
  • Withstand temperatures to 1150 °C

To prevent thermal stress cracks, we strongly recommend a heating/cooling rate not exceeding 200 °C per hour. United porcelain crucibles offer excellent resistance to acids and alkalies except for hydrofluoric acid.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty