Would you like to log in again?
Transport Tube, Sterile
Supplier: United Scientific Supplies
UNG1028
470339-688EA 223.5 USD InStock
470339-688
Transport Tube, Sterile
Tubes Test Tubes
Sterilized polystyrene transport tubes with polyethylene screw cap.
  • Pack of 500 sterilized polystyrene transport tubes with 8ml capacity
  • Self-standing design
  • Include leak-proof polyethylene screw caps
  • 20 mm diameter×112 mm height (with cap)
Pack of 500 sterilized polystyrene transport tubes with 8 ml capacity. Feature raised graduations from 1 to 8 ml with graduation intervals of 1 ml. Designed with flat bottom. Leak-proof polyethylene screw caps included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty