Would you like to log in again?
Equal Mass Density Cylinders, Metals
Supplier: United Scientific Supplies
SGMC05
470339-692EA 28.3 USD InStock
470339-692
Equal Mass Density Cylinders, Metals
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Equal mass cylinders for density investigations.
  • Set of 5 cylinders made of diverse materials, each with identical mass and radius
  • Ideal for lessons around specific gravity
  • Storage box and activity guide also included
Each specimen in this set of five diverse materials cylinders has identical mass and radius. Materials included are aluminum, zinc, tin, steel, and copper. Includes a storage box and activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty