Would you like to log in again?
United Scientific™ Lumi Rod (Light Pipe)
Supplier: United Scientific Supplies
LUMRD48 LUMRD24
470339-468EA 28 USD InStock
470339-468 470339-706
United Scientific™ Lumi Rod (Light Pipe)
Educational Materials Physics Educational Materials Optics Learning Activities
Demonstrate the internal reflection of a laser.
  • Excellent tool for demonstrating the principle of fiber optics
  • Curled clear acrylic rod conducts light by internal reflection
  • Works with flashlight or laser pointer
  • Includes activity guide
Excellent tool for demonstrating the principle of fiber optics. The curled clear acrylic rod conducts light by internal reflection. A flashlight or laser pointer can be used for the demonstration.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty