Would you like to log in again?
3-Prong Clamp with Flexible Extension Rod
Supplier: United Scientific Supplies
UNFLEXCLP01
470339-746EA 63.65 USD InStock
470339-746
3-Prong Clamp with Flexible Extension Rod
Clamps General Purpose Clamps
Sturdy flexible support arm with clamp.
  • Flexible arm
  • Holds up to 2 pounds
This flexible arm is extremely versatile and can be placed in virtually any position or angle. Includes a 3-prong clamp head, and nickel-plated flex arm 305 mm (12") in length. The Lab-Frame Connector securely attaches to support stands, lab-frames, or any support rod up to 19 mm (0.75") diameter.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty