Would you like to log in again?
Protozoa and Algae Slide Set Set of 20
Supplier: United Scientific Supplies
UNSLPRAL20
470339-754EA 75.15 USD InStock
470339-754
Protozoa and Algae Slide Set Set of 20
Slides Prepared Slides
Collection of popular Protozoa and Algae samples.
  • Glass prepared microscope slides
  • Plastic storage case
  • Key chart is included on lid
Glass prepared microscope slides, supplied in a plastic case. A collection of slides specific to the study of protozoa and algae. Set of 20.
Set includes: 1) Amoeba, E, 2) Mixed foraminifera, 3) Mixed radiolaria, 4) Euglena, E, 5) Trypanosoma in blood smear, 6) Plasmodium sp. (Malaria) in human blood, 7) Paramecium, E, 8) Paramecium showing conjugation, 9) Vorticella, E, 10) Chlamydomonas, E, 11) Pandorina, E, 12) Volvox, with daughter colonies, E, 13) Nostoc, filaments with heterocysts, E, 14) Cladophora, E, 15) Chlorella, E, 16) Spirogyra, vegetative filaments, E, 17) Spirogyra, conjugation, E, 18) Desmids, mixed species, E, 19) Diatoms, mixed species, E, 20) Paramecium fission.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty