Would you like to log in again?
Electric Bell Model
Supplier: United Scientific Supplies
ELBM01
470339-770EA 23.8 USD InStock
470339-770
Electric Bell Model
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate how electric coils can be used to power a bell.
  • Electric bell model mounted on a clear acrylic plate and finished wooden stand
  • On/off switch, adjustable contact screw, and binding posts for easy connection
  • Operates on 6 V DC
  • Includes activity guide
This well constructed and attractive electric bell model is mounted on a clear acrylic plate and a finished wooden stand to allow the functional parts to be clearly seen. Model operated on 6 V DC with an on/off switch, adjustable contact screw, and binding posts for easy connection. Includes activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty