Would you like to log in again?
Dual Color Laser Pointer
Supplier: United Scientific Supplies
DLP006
470339-808EA 75.6 USD InStock
470339-808
Dual Color Laser Pointer
Laser Pointers
Dual red and green laser pointer for optical experiments.
  • Class 3R (Class IIIa) laser <5 mW
  • Green wavelength of 532 nm (±10 nm)
  • Red wavelength of 650 nm (±10 nm)
  • Expected operating life is 1000 hours
Economical laser pointers are useful for many optical experiments. Lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip. The lasers are Class 3R, and are each L×D: 6×0.5".
Red laser pointer has a maximum output of 5 mW and a wavelength of 650 nm (±10 nm). Range of beam is 100 meters. Expected operating life is 1000 hours. Operates on two 'AAA' batteries (not included).
Green laser pointer has a maximum output of 5 mW and a wavelength of 532 nm (±10nm). Range of beam is 200 meters. Expected operating life is 1000 hours. Operates on two 'AAA' batteries (not included).
Dual laser pointer (red/green) has a maximum output of 2×5 mW and a green wavelength of 532 nm (±10 nm) and red wavelength of 650 nm (±10 nm). Range of green beam is 200 meters. Range of red beam is 100 meters. Expected operating life is 1000 hours. Operates on two 'AAA' batteries (not included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty