Would you like to log in again?
Mortar and Pestle Set, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
JMD275 JMD050 JMD400 JMD150 JMD070 JMD1900 JMD750
470339-254EA 28.6 USD InStock
470339-254 470339-596 470339-372 470339-296 470340-276 470339-846 470340-046
Mortar and Pestle Set, Porcelain, United Scientific Supplies
Mortars and Pestles Mortars with Pestles
Deep form mortar with oversized pestle.
  • Mortar features pour spout and is glazed outside (excluding bottom), unglazed inside
  • Oversized pestle is glazed to grinding surface
Deep-form mortar with pour spout. Glazed on the outside (except for the bottom) and unglazed inside. Oversized pestle is glazed to grinding surface.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty