Would you like to log in again?
Deluxe Metalware Kit
Supplier: United Scientific Supplies
UNMTLK-1
470339-936EA 35.7 USD InStock
470339-936
Deluxe Metalware Kit
Support Stands
This deluxe set includes a support stand with a cast iron base (5×8") with steel support rod (20"), steel support ring (3"), a 2-prong extension clamp with cork lined jaws, and an aluminum clamp holder. The support rod includes a 3" long rod to provide more torque when securing the rod to the base plate.
  • Sturdy metal components
  • Iron base and steel supports
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty