Would you like to log in again?
United Scientific™ Overflow Can And Catch Bucket Set
Supplier: United Scientific Supplies
OFLSET
470339-984EA 19.25 USD InStock
470339-984
United Scientific™ Overflow Can And Catch Bucket Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
Aluminum catch can for a variety of experiments.
  • Aluminum
  • For use in Archimedes' Principle, density, and specific gravity experiments
An aluminum overflow can and a catch bucket are packaged as a convenient set for use in Archimedes' Principle, density, and specific gravity experiments. The overflow can measures 4ID×5" high and has a capacity of approximately 800 ml. Its spout is 0.5ID×1.5" long. The catch bucket with detachable aluminum handle measures 4×3" high.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty