Would you like to log in again?
Demo Diffraction Grating
Supplier: United Scientific Supplies
DFG003
470339-988EA 34.65 USD InStock
470339-988
Demo Diffraction Grating
Educational Materials Physics Educational Materials Optics Learning Activities
Convenient diffraction grating with three different spacings
  • Demonstration slide features three 20×10 mm gratings with 100, 300 and 600 lines per mm
  • Mounted between glass plates
  • Viewing area of each section is 15×10 mm
Use this diffraction grating to demonstrate the optical properties of three different diffraction gratings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty