Would you like to log in again?
Miniature DC Motor for Solar Experiments
Supplier: United Scientific Supplies
DCM015-C
470339-998EA 3.85 USD InStock
470339-998
Miniature DC Motor for Solar Experiments
Educational Materials Physics Educational Materials Mechanics Learning Activities
A perfect motor for solar cell experiments.
  • Solar motor specifically designed for experiments using low starting voltage (0.5 V) and current (25 mA)
  • Speed: 1000 to 5000 rpm
  • Includes soldered-on 4" wire leads
Motor is approximately 38 mm long×24 mm in diameter. Shaft size is 8 mm long×2 mm in diameter. Includes soldered-on 4" wire leads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty