Would you like to log in again?
Labclean™ Cleanroom Wipes, 55% Cellulose/45% Polyester, United Scientific Supplies
Supplier: United Scientific Supplies
CPWIPE9X9 CPWIPE4X4 CPWIPE12X12
470339-888EA 91.8 USD InStock
470339-888 470340-100 470340-002
Labclean™ Cleanroom Wipes, 55% Cellulose/45% Polyester, United Scientific Supplies
Wipes Cleanroom Wipes Dry Cleanroom Wipes
100% polyester wipes for use in cleanroom environments.
  • 100% virgin, continuous filament, double-knit polyester fiber
  • Laser cut, sealed edge for reduced fiber contamination
  • Moderate absorbency and abrasion resistance
  • Low levels of particulate and extractable counts
Spunlace non-woven wipes are constructed from a hydro-entangled, non-woven blend of 55% cellulose and 45% polyester. This composite substrate has the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic yarn. Recommended for use in cleanroom Class 10000+ (ISO 7+) critical environments such as research labs, hospitals, and electronics manufacturers. Laundered and packaged in a cleanroom. Vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with an inner bag label. Available in three convenient sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty