Would you like to log in again?
Thermoelectric Demonstrator
TED001
470340-112EA 6.69 USD InStock
470340-112
Thermoelectric Demonstrator
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Demonstrate how the Second Law of Thermodynamics can create electricity.
  • Used to demonstrate the Second Law of Thermodynamics
  • Includes a set of two 24" banana connecting cords and a detailed activity guide
Demonstrate the Second Law of Thermodynamics with this apparatus. Immerse one of the aluminum legs into ice cold water and the other leg into hot water to demonstrate how heat transfer can be converted to electrical energy. This conversion turns the turbine. Show the reverse effect of converting electrical energy to heat energy by plugging the apparatus into a DC power supply. In this activity, DC current is given to the unit so one of the strips will heat up and the other will remain cold.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty