Would you like to log in again?
Jade Bottle Top Dispenser, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BTJD2.5 BTJD5 BTJD100 BTJD60 BTJD30 BTJD10
470339-930EA 683.4 USD InStock
470339-930 470339-442 470339-774 470339-972 470339-670 470340-144
Jade Bottle Top Dispenser, United Scientific Supplies
Bottle Top Dispensers
Precise dispensing for a wide range of reagents.
  • Fully autoclavable bottle top dispenser
  • Includes adapters to fit a wide range of bottle sizes: 28, 32, 38, 40 and 45 mm
  • Features re-circulation valve to redirect liquid back into the bottle without reagent loss
  • Smooth plunger movement enables bubble-free dispensing
Five adapters are included with each unit for use with a wide range of bottle sizes. Adapters included are: 28, 32, 38 , 40 and 45 mm.
Fully autoclavable dispenser features a re-circulation valve that redirects liquid back into the bottle without the loss of reagent. Operates with a smooth plunger movement for bubble-free dispensing and effortless performance, even under demanding laboratory conditions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty