Would you like to log in again?
Micro Centrifuge Tubes, PP, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
P10201A P10204 P10204R P10205 P10203A P10201 P10202 P10207 P10203 P10204A
470339-700EA 19.7 USD InStock
470339-700 470339-782 470339-794 470339-222 470339-806 470340-178 470339-648 470339-418 470338-868 470339-040
Micro Centrifuge Tubes, PP, United Scientific Supplies
Tubes Microcentrifuge Tubes
High-quality microcentrifuge tubes for safer use in molecular biology and other biochemical applications.
  • Made from high-quality virgin polypropylene, RNase, DNase and Pyrogen free
  • Feature large, molded-in graduations and frosted surface for easy writing
  • Can be used in very low temperatures, autoclavable
  • Maximum RCF is 20000
Made from high-quality virgin polypropylene, these tubes are RNase, DNase and Pyrogen free for safer use in molecular biology and other biochemical applications. Feature large molded-in graduations and a frosted surface for easier writing. Autoclavable tubes can be used in very low temperatures and with various organic solvents.
Color: Natural
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty