Would you like to log in again?
Magnetic Field Demonstrator
Supplier: United Scientific Supplies
MGFD03
470340-218EA 43.25 USD InStock
470340-218
Magnetic Field Demonstrator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Large viewer to illustrate magnetic fields.
  • Quick and clean tool to visualize magnetic fields
This is a quick and clean way to visualize magnetic fields. The demonstrator is a transparent acrylic case that holds iron filings suspended in a fluid of mineral oil. When it is placed over a magnet, the field pattern can be viewed directly, or it can be placed on an overhead projector for classroom demonstration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty