Would you like to log in again?
United Scientific Aluminum Alloy Tuning Fork Sets
Supplier: United Scientific Supplies
TFSET4 TFSET13 TFSET8
470339-640EA 43.05 USD InStock
470339-640 470340-272 470353-000
United Scientific Aluminum Alloy Tuning Fork Sets
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Aluminum tuning fork sets.
  • Made of aluminum alloy
Convenient assortment of eight tuning forks. Contains one each of the following frequencies: 256, 288, 320, 341.3, 384, 426.6, 480, and 512 Hz. Designed with frequency and scale letter stamped on each fork.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty