Would you like to log in again?
Mortar and Pestle Set, Agate Stone, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MPA060 MPA015 MPA035 MPA040 MPA030 MPA020
470339-402EA 115.1 USD InStock
470339-402 470340-278 470339-184 470339-360 470339-044 470339-172
Mortar and Pestle Set, Agate Stone, United Scientific Supplies
Mortars and Pestles Mortars with Pestles
Natural agate stone mortar and pestle.
  • Octagonal mortar and pestle set
  • Crafted from naturally hard agate stone, with highly polished grinding surfaces
  • Surface patterns will vary
  • Ideal for use in laboratories demanding contamination-free results
Octagonal mortar and pestle sets are made of naturally hard agate stone (6.5 on Mohs' Scale), with highly polished grinding surfaces. Ideal for use in laboratories demanding contamination-free results. Note that the natural agate material causes surface pattern variations in finished mortars and pestles.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty