Would you like to log in again?
IV Tubing Sets
Supplier: MEDLINE INDUSTRIES, INC.
DYNDTN1539 DYNDTN1545 DYNDTN0512 DYNDTN1512B DYNDST1545 DYNDTA1540 DYNDTN1529 DYNDTN1537 DYNDTN1512 DYNDST1512
470340-424CS 444.59 USD InStock
470340-424 470340-366 470340-388 470340-400 470340-416 470340-428 470340-394 470340-418 470340-398 470340-420
IV Tubing Sets
Infusions
Soft tubing for patient comfort.
  • Designed for gravity use only
  • IV tubing is not compatible with infusion pump administration systems
  • Available with up to 3 ports
Infuse medications and fluids accurately and consistently with Medline IV sets and accessories. Available with different components and configurations to accommodate a range of clinical applications, our IV products offer you clinical benefits and superior quality.
Certifications: Does not contain DHEP.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty