Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes Star Hopper Model Rockets

Supplier: ESTES INDUSTRIES INC SE
Estes
7303 1721
470343-086PK 159.9 USD InStock
470343-086 470343-092
Estes Star Hopper Model Rockets
Educational Materials Physics Educational Materials Mechanics Learning Activities
Easy-to-assemble beginner level rocket. No tools or glue needed.

  • Beginner-level build
  • Requires mini size 13 mm 1/2A, A3, or A10 rockets
  • Max. altitude 122 m (400')

An amazingly cool looking beginner model rocket that doesn't require any glue or tools to build! Estes' snap-together star hopper rocket shines with its retro atomic-age silver paint scheme and detail-molded plastic fins, nose cone, and engine mount. A long, 18" streamer provides an efficient and safe recovery system. Star hopper is the answer for cub scouts and other younger rocketeers. They can have the fun of building without the frustration of dripping glue and aligning fins! Star Hooper assures your kids have a first class model rocketry experience.

Ordering information: Launch system and rocket engines available separately. Single kit includes all parts needed to assemble (1) model rocket. No tools/glue needed. Bulk pack includes: (12) single kits.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty