Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes Viking Model Rockets

Supplier: ESTES INDUSTRIES INC SE
Estes
1949 1755
470343-074EA 20.25 USD InStock
470343-074 470343-088
Estes Viking Model Rockets
Educational Materials Physics Educational Materials Mechanics Learning Activities
Intermediate-level rocket with five fiberboard fins and a streamer recovery system.

  • Intermediate-level build
  • Requires standard size 18 mm A, B, or C engines (A or B recommended)
  • Max. altitude 488 m (1600')

Get ready to launch your creativity to new heights with the Estes Viking model rocket! The very versatile Viking is everyone's favorite research rocket to use with classroom investigations. This rocket has 48 possible fin configurations that can be used for it. Students can hypothesize how their fin arrangement will affect the rocket’s flight then put it to the test by launching it. The Viking is the perfect, low cost rocket for slim budgets.

Level-up your rocket assembly and finishing skills with the Viking's five lightweight fiberboard fins. The Viking can soar to heights of up to 1600 feet before gently returning to earth on its brightly colored parachute. The Viking is an exciting and challenging model rocket for budding scienitists, engineers, and hobbyists.

Ordering information: Launch system and rocket engines available separately. Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required. Bulk pack includes: (12) single kits.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty