Would you like to log in again?
Estes Altimeter
Supplier: ESTES INDUSTRIES INC SE
Estes
2246
470343-102EA 59.3 USD InStock
470343-102
Estes Altimeter
Educational Materials Physics Educational Materials Mechanics Learning Activities
Add some tech power to your rocket experiments and take the guesswork out of altitude measurements.
  • Record up to 10 flights at a time
  • Display altitude in either meters or feet
  • Max. altitude 3000+ m (9999')
Half the fun of model rockets is building them, half the fun is launching them, and the other half analyzing the data! No rocketry lesson is complete unless your rocketeers can answer the quintessential question: 'How high did it go?' The Estes digital altimeter is an exciting, easy way to take your rocketry lesson to new heights, with real, accurate altitude data to analyze after the launch.
Small enough to be used in model rockets only 20 mm in Ø, the easy to read LCD display can be toggled between launches and units. Cut or drill four small holes in the body tube of your rocket to allow accurate pressure readings.
Ordering information: Please note that the Estes altimeter may only be used in rockets with a body tube Ø of at least 20 mm. Compatible models include Alpha, Alpha III, Generic E2X, green eggs and more. The Gnome, Viking, Wizard and more are too small.
Delivery information: Includes replaceble 6 V alkaline battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty