Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes Gnome Model Rockets

Supplier: ESTES INDUSTRIES INC SE
Estes
886 1749
470343-070PK 88.4 USD InStock
470343-070 470343-112
Estes Gnome Model Rockets
Educational Materials Physics Educational Materials Mechanics Learning Activities
Easy-to-assemble beginner level rocket with pre-colored parts.

  • Beginner-level build
  • Requires mini size 13 mm 1/4A, 1/2A, A3, or A10 engines
  • Max. altitude 244 m (800')

Looking for a mini engine powered rocket that's as stylish as it is high-flying? The Estes Gnome is chrome at its hottest! Standing just a little over 10 inches tall with super shiny chrome and sapphire blue colors, this is one great looking, mini engine powered cloud chaser that can reach altitudes of 800 feet.

The Gnome is a beginner skill level kit that's easy to assemble and comes with a chrome wrapped body tube, a pre-colored one-piece fin unit, nose cone, and upper launch lug ring. Once assembled, the rocket is finished off with a colorful self-stick decal. Recovery is fast and reliable thanks to the orange colored streamer, making it easy to track the Gnome after a super-fast flight.

Ordering information: Launch system and rocket engines available separately. Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required. Bulk pack includes: (12) single kits.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty