Would you like to log in again?
Estes Green Eggs Model Rockets
Supplier: ESTES INDUSTRIES INC SE
Estes
1718 7301
470343-084EA 26.5 USD InStock
470343-084 470343-118
Estes Green Eggs Model Rockets
Educational Materials Physics Educational Materials Mechanics Learning Activities
Launch an egg and test recovery systems with this reliable, sturdy model rocket.
  • Intermediate-level build
  • Large rocket can accommodate a chicken egg payload
  • Requires large size 24 mm C11 or D engines
Looking for an exciting and educational way to teach your students about rocket science and engineering? Look no further than the Estes Green Eggs model rocket. This kit is specifically designed for school science projects and STEM classes, and is the perfect way to introduce your students to the world of rocketry.
Designed for Estes C11 and D engines, this rocket model can safely carry a regular-sized raw egg as its payload. With its clear payload section, you can even fit two eggs with a D12-3 engine! The Green Eggs rocket's swept fins not only add stability during flight but also allow the Green Eggs rocket to stand by itself. Additionally, the balsa fins are thick enough to enhance alignment and provide exceptional strength, making it suitable even for mounting through a wall. With a main body tube and clear payload section of the same diameter, the green eggs rocket is easy to set up with a standard launch lug without the need for awkward stand-offs.
Overall, the Green Eggs lofting rocket provides a safe and fun way for your students to conduct experiments with their "eggstronauts" without spending too much time on building. Take your "egg drop" experiments to new heights, and engage your students in engineering design.
Ordering information: Launch system and rocket engines available separately.
Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required.
Bulk pack includes: (12) single kits.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty