Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes Wizard Model Rockets

Supplier: ESTES INDUSTRIES INC SE
Estes
1292 1754
470343-120EA 16.55 USD InStock
470343-120 470343-068
Estes Wizard Model Rockets
Educational Materials Physics Educational Materials Mechanics Learning Activities
Intermediate-level rocket with laser-cut balsa fins and streamer recovery system.

  • Intermediate-level build
  • Requires standard size 18 mm A, B, or C engines (A or B recommended)
  • Max. altitude 488 m (1600')

Looking for a magical experience? Are your model rocketry skills ready to level-up? The Estes wizard rocket is exactly what you need! This high-performance rocket is easy to build and known for casting spells on all who own it.

With powerful Estes model rocket engines, this light-weight rocket can reach heights of up to 1600 feet in the blink of an eye. Its laser-cut balsa wood fins still make assembly easy, and its streamer recovery system helps it descend efficiently and safely from high altitudes. This sleek rocket is fun to build and thrilling to fly.

Ordering information: Launch system and rocket engines available separately.

Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required.

Bulk pack includes: (12) single kits.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty