Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes Alpha Model Rockets

Supplier: ESTES INDUSTRIES INC SE
Estes
1756 1225
470343-082EA 28.25 USD InStock
470343-082 470343-124
Estes Alpha Model Rockets
Educational Materials Physics Educational Materials Mechanics Learning Activities
Easy-to-assemble intermediate-level rocket with laser cut balsa fins.

  • Intermediate-level build
  • Requires standard size 18 mm A, B, or C engines
  • Max. altitude 305 m (1000')

The Estes alpha is a rocket that has stood the test of time and continues to be the go-to choice for educators after 40 years. Designed to be easy to build and customize, it offers a flexible platform for teaching valuable STEM principles to students. The unforgettable experience of launching an Estes alpha rocket is something that stays with people for a lifetime, with millions of individuals remembering their first launch with fondness.

With its easy assembly and reliable performance, the Alpha is sure to hook any student who gives it a try. Not only is Alpha easy to build and fly, it also looks great with its suggested color scheme and red, white, and blue decals. And with the ability to reach cloud-topping altitudes of 1000 feet, this rocket is sure to provide hours of entertainment.

Ordering information: Launch system and rocket engines available separately.

Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required.

Bulk pack includes: (12) single kits.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty