Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes AVG Model Rockets

Supplier: ESTES INDUSTRIES INC SE
Estes
1753
470343-128PK 110.25 USD InStock
470343-128
Estes AVG Model Rockets
Educational Materials Physics Educational Materials Mechanics Learning Activities
Alpha, viking and generic E2X rockets in one education bulk pack.

  • Intermediate-level build
  • Requires standard size 18 mm A, B, or C engines
  • Max. altitude 488 m (1600')

Get ready to launch into the world of rocketry with the AVG bulk pack! This complete set includes rockets from three of our popular bulk packs, providing teachers and group leaders with a variety of skill levels and rocket types in a single package.

The pack includes 4 each of the classic intermediate level alpha, 4 each of the versatile intermediate level Viking with 48 fin combinations, and 4 each of the beginner level generic E2X. Each model requires a few basic tools and supplies for building, such as scissors, sandpaper, glue, and paint.

This all-in-one rocket pack is perfect for classroom and club settings and offers a range of rocket-building and launching experiences for beginners and intermediate-level hobbyists alike.

Ordering information: Launch system and rocket engines available separately.

Bulk pack includes: (4) Alpha model rocket kits, (4) Viking model rocket kits, and (4) Generic E2X model rocket kits. Some tools and glue may be required.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty