Would you like to log in again?
Adjustable Boss Head Clamp
Supplier: GSC INTERNATIONAL SE
4-ACH40-10
470343-240PK 107.35 USD InStock
470343-240
Adjustable Boss Head Clamp
Clamps Bossheads
These boss head clamp holders are adjustable up to 16 mm.
  • Accepts rods up to 5/8" in diameter
  • Made with cast-iron body and nickel-plated thumb-screws
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty