Would you like to log in again?
Innovating Science® Presumptive Gun Shot Residue Kit
Supplier: ALDON CORP SE
IS9001
470343-392EA 56.6 USD InStock
470343-392
Innovating Science® Presumptive Gun Shot Residue Kit
Educational Classroom Kits and Activities
Learn about how to test if a surface has been exposed to gunpowder residue.
  • Students will perform a two part test to determine whether a surface has been exposed to a discharged firearm
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
This is a two part test to determine whether a surface has been exposed to a discharged firearm. A rapid color change takes place to verify the presence of nitrates and lead. Kit contains instructions and enough materials for 30 tests.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, Lead and lead compounds, which are known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty