Would you like to log in again?
Innovating Science® Presumptive Blood Test Kit
Supplier: ALDON CORP SE
IS9002
470343-394EA 34.6 USD InStock
470343-394
Innovating Science® Presumptive Blood Test Kit
Educational Classroom Kits and Activities
Learn about how to test for the presence of blood on materials.
  • Students will test materials for the presence of blood.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-3
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
Test for the presence of blood on materials using phenolphthalein. The test will not distinguish between animal and human blood. Further serology tests are needed. Kit contains instructions, blood standard and reagents to complete 30 tests.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Phenolphthalein and Methanol, which are known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty