Would you like to log in again?
Innovating Science® Chemiluminescence
Supplier: ALDON CORP SE
IS2502
470343-430EA 66.3 USD InStock
470343-430
Innovating Science® Chemiluminescence
Educational Classroom Kits and Activities
Learn about light energy and light-emitting reactions called chemiluminescence.
  • Students will create a chemiluminescent reaction and observe the blue glow it creates.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2, HS-PS1-4, HS-PS3-3
  • Disciplinary core ideas: PS1.A, PS1.B, PS3.A
  • Cross cutting concepts include energy and matter, structure and function, patterns
  • Engineering practices include developing and using models, analyzing and interpreting data, obtaining evaluating and communicating information
Most physical and chemical reactions that involve the release of light energy also involve the release of heat energy. One unique and interesting form of light-emitting reaction is called chemiluminescence, or 'cool light'.
Similar to the familiar glow of a firefly, students will create a chemiluminescent reaction in the classroom, and then observe the fascinating blue glow of the reaction, which lasts for over ten minutes. This is chemistry that never fails to amaze students. The kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Kit includes instructional manual and SDS booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty