Would you like to log in again?
Innovating Science® Organic Chemistry: Chemical Identification of Biomolecules
Supplier: ALDON CORP SE
IS2508
470343-440EA 68.35 USD InStock
470343-440
Innovating Science® Organic Chemistry: Chemical Identification of Biomolecules
Educational Classroom Kits and Activities
Learn about the importance of proteins, carbohydrates, and lipids in living organisms.
  • Students will learn to test for presence of proteins, sugars, starch, and lipids.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
Understand the importance of proteins, carbohydrates, and lipids in living organisms. Students will learn to identify a positive test result for proteins using biuret reagent, examine the reaction between Benedict's reagent and a simple sugar, use iodine/potassium iodide to test for the presence of starch, and test for the presence of lipids using a fat-soluble dye. Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Kit includes instructional manual and SDS booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty