Would you like to log in again?
Innovating Science® ATP Muscle Kit
Supplier: ALDON CORP SE
IS3015
470343-494EA 185 USD InStock
470343-494
Innovating Science® ATP Muscle Kit
Educational Classroom Kits and Activities
Learn about muscles and how/why they contract.
  • Students will observe and measure how muscles contract using an ATP solution
  • Aligned to NGSS Practices
  • Performance expectations: HS-LS1-2
  • Disciplinary core ideas: LS1.A
  • Cross cutting concepts include systems and system models
  • Engineering practices include developing and using models
Students can observe and measure how muscles contract. The muscles are shipped in glycerol which extracts most of the water soluble material leaving muscle tissue that can be identified and studied using a microscope. The muscle can also be stimulated to contract using the ATP chemical solution. There are enough materials for 15 groups.
Delivery information: Kit includes instructional manual and SDS booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty