Would you like to log in again?
Innovating Science® Plant Food - Nutrient Deficiency in Plants
Supplier: ALDON CORP SE
IS3050
470343-498EA 76.45 USD InStock
470343-498
Innovating Science® Plant Food - Nutrient Deficiency in Plants
Educational Classroom Kits and Activities
Learn the effects of nutrient deficiency in plants.
  • Students will deprive plants of prepared nutrient solutions to determine the effects of specific nutrients on plant growth and appearance.
  • Aligned to NGSS Practices
  • Performance expectations: MS-LS2-1
  • Disciplinary core ideas: LS2.A
  • Cross cutting concepts include cause and effect
  • Engineering practices include analyzing and interpreting data
Using the materials provided, students will be able to examine the effects of nutrient deficiency on plants. Specially prepared nutrient solutions (included in the kit) will allow students to deprive each plant of one of seven specific vital nutrients. Over time, students will determine the effects, if any, of the nutrient deprivation through physical observation of the plant's growth.
Kit contains enough materials for three complete set-ups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Kit includes instructional manual and SDS booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty