Would you like to log in again?
Innovating Science® Coliform Test Powder Kits
Supplier: ALDON CORP SE
Innovating Science®
IS5055 IS5056
470343-536EA 42.29 USD InStock
470343-536 470343-534
Innovating Science® Coliform Test Powder Kits
Media for Microbiology Dehydrated Media for Microbiology
Easy way to test water for coliforms.
  • One water test for presence of coliforms
Simply add a small amount of collected water to the sample tube and incubate for 24 to 48 hours. A distinct color change from purple to yellow indicates that coliforms are present.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty