Would you like to log in again?
Eisco Double Convex Glass Lenses
Supplier: EISCO SCIENTIFIC LLC
Eisco
PH0534ECV PH0534OCV PH0534RCV PH0534RCV2 PH0534FCV PH0534KCV PH0534GCV PH0534HCV PH0534CCV PH0534DCV PH0534NCV PH0534PCV PH0534RCV1 PH0534BCV PH0534JCV PH0534ACV PH0534ICV PH0534QCV
470328-068EA 6.3 USD InStock
470328-068 470344-090 470344-182 470345-708 470344-354 470344-550 470344-644 470344-116 470344-646 470345-032 470328-070 470343-600 470328-072 470345-396 470345-212 470328-074 470345-002 470328-066
Eisco Double Convex Glass Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
Individual spherical glass lenses.
  • Double convex lenses in a variety of diameters and focal lengths
  • Made of high quality, optically worked glass
  • Highly polished surface, ground edges
This optically true lens is excellent for physics classroom demonstrations in optics and light refraction.
Eisco labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments. These lenses are designed for educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty