Would you like to log in again?
Eisco T-Shaped Nesting Retort Bases, Cast Iron
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Eisco
CH0656H CH0656G
470343-606EA 29.75 USD InStock
470343-606 470344-976
Eisco T-Shaped Nesting Retort Bases, Cast Iron
Support Stands
T-Shaped Retort Bases designed to nest closely to each other, made of chemically-resistant cast iron.
  • T-shaped retort bases made of chemically-resistant cast iron with a black coating
  • Designed with one raised leg that allow multipe bases to slide freely under one another for compact setup or storage
  • Heavy and sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
T-Shaped Retort Stands are made of chemically-resistant cast iron with a black coating. One raised leg allows multipe bases to slide freely under one another for compact setup or storage. Features a tapped hole approximately ¹/₃ of the way from the intersection of the legs, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for both science classrooms and professional labs.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty