Would you like to log in again?
Polypropylene Test Tube Rack
Supplier: EISCO SCIENTIFIC LLC
CH0694A
470343-622EA 2.29 USD InStock
470343-622
Polypropylene Test Tube Rack
Racks Tube Racks
This rack is made of durable polypropylene and securely holds test tubes with diameters of 30, 20, 17 and 12 mm for organized laboratory storage.
  • Test tube rack featuring a sturdy polypropylene construction that will stand up to the wear and tear of the lab. Comes in blue or white color
  • Supports up to 6 test tubes, with 2 holes that measure up to 25 mm in diameter and 4 holes that measure up to 16 mm in diameter - Provides convenient insertion and removal of tubes
  • Rack has 6 vertical pins for drying tubes
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Polypropylene test tube rack is complete with 6 drying pins, 10 mm diameter, and 6 holes. The two outer holes will accommodate tubes up to 1" in diameter. The 4 inner holes will accommodate tubes up to 0.75" in diameter. It measures 10" in length and 3" in height and is fully autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty