Would you like to log in again?
Eisco High-Performance Variable Volume Micropipettes
Supplier: EISCO SCIENTIFIC LLC
VVM5 VVM8 VVM2 VVM13 VVM7 VVM4 VVM10 VVM11 VVM1 VVM9 VVM12 VVM6 VVM3
470343-816EA 136.5 USD InStock
470343-816 470345-910 470344-184 470343-726 470343-626 470344-014 470344-602 470344-854 470345-692 470345-790 470343-810 470345-100 470345-366
Eisco High-Performance Variable Volume Micropipettes
Pipettes Micropipettes
Variable volume micropipettes are fully autoclavable and feature a simple ergonomic design for comfort, color coded plungers, and calibration conforming to DIN ISO standards.
  • Soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use
  • Fully autoclavable with a durable, autoclavable PVDF tip cone
  • Slip-resistant thermoplastic elastomer (TPE) body grip to minimize body heat transfer to the micropipette during long use, resulting in higher accuracy
  • Color coded plungers for easy identification
  • Calibration conforms to DIN 12650 & EN-ISO 8655 standard ensuring high accuracy and precision - Calibration report enclosed with every micropipette
High performance variable volume micropipettes have a simple design which allows for easy adjustment and reading of increments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty