Would you like to log in again?
Eisco Burette Stopcock Plunger (PTFE)
Supplier: EISCO SCIENTIFIC LLC
CH0233BSPR
470343-680EA 9.75 USD InStock
470343-680
Eisco Burette Stopcock Plunger (PTFE)
Burets
Replacement stopcock for Eisco acrylic burette, part number 470343-708.
Replacement stopcock for Eisco item number CH0233B. Features three rubber gaskets that connect to the burette. Handle spins in a full circle to adjust the flow rate of the stopcock. Measures 3⁵/₁₆" from end to end. The top measures ⁷/₁₆" diameter. Tip is approximately ¹/₈" with a ¹/₁₆" diameter hole.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty