Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Individual Brass Test Sieves, 8" Full Height, ASTM

Supplier: EISCO SCIENTIFIC LLC
BI0139BRE BI0139BRB BI0139BRF BI0139BRC BI0139BRA
470346-004EA 42.7 USD InStock
470346-004 470343-706 470344-870 470345-718 470345-246
Eisco Individual Brass Test Sieves, 8" Full Height, ASTM
Sieves Test Sieves
Stackable premium quality test sieves for use with many different particle-sizing applications.

  • Features stainless steel woven-wire cloth
  • Sieve is full height 2.5" and measures 203 mm (8") in Ø

Made of high quality brass and stainless steel - built to last and withstand multiple uses.

Certifications: Meets ASTM E11 and AASHTO M 92 requirements for compliance test sieves.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty