Would you like to log in again?
Eisco Acrylic Burettes with PTFE Key Stopcock, Class B
Supplier: EISCO SCIENTIFIC LLC
CH0233A CH0233B
470343-708EA 24.89 USD InStock
470343-708 470344-842
Eisco Acrylic Burettes with PTFE Key Stopcock, Class B
Burets
Lightweight and durable.
  • Made of high quality acrylic; tolerates most dilute minerals, acids, and bases
  • Class B, ±0.10 ml tolerance as per DIN ISO 385 standards for precision
  • Chemical-resistant blue printed scale in 0.10 ml graduations
  • Includes an attached PTFE straight bore stopcock
This acrylic burette resists shattering, impact, and most non-abrasive cleaners including ammonia and strong detergents. The ±0.10 ml tolerance meets ASTM Class B / DIN ISO 385 standards for precision. Comes fitted with a polytetrafluoroethylene (PTFE) plastic plunger key stopcock affording low friction use while resisting chemicals, moisture, and high temperatures. The burette has an easy-to-read blue scale with 0.10 ml graduations. Calibrated for delivery. Note: Not for use with hydrofluoric acid or ammonium hydroxide. Not autoclavable.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty